Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ HCV Seroconversion Panel, Modified

Catalog No. 22556289 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
9 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-556-289 9 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-556-289 Supplier SeraCare™ Supplier No. 06100218
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Consists of undiluted aliquots from a single individual's plasma units, genotype 1a

  • Collected over 35 days from 1999 to 2000
  • No preservatives added
  • Some panel members were found positive for anti-HCV; All were found negative for HBsAg and anti-HIV
  • For research use only; Not for diagnostic procedures

Specifications

Content And Storage -65°C to -80°C
Format Panel
Type Hepatitis C Seroconversion Panel
Includes Set of nine undiluted Plasma samples from a single Plasma donor collected during a period of HCV seroconversion
Intended Use Evaluating HCV test system using well characterized specimens, to provide comprehensive data for comparative analysis
Quantity 9 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.