Learn More
SeraCare™ Hepatitis B Seroconversion Panels
Catalog No. 22554534 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
6 x 1.5 mL
8 x 1.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-534 6 x 1.5 mL
22-554-535 8 x 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-554-534 Supplier SeraCare™ Supplier No. 06050026
Add to Cart
Add to Cart
Undiluted plasma samples collected from a single donor during development of infection and subsequent immune response
For research use only.
Specifications
Format Panel
Type Hepatitis B Seroconversion Panel
Intended Use For evaluating HBV test system using well characterized specimens, to provide comprehensive data for comparative analysis
Quantity 6 x 1.5 mL