Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ HIV 1/2/O/p24 Qualification-Panel

Catalog No. 21123108 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
6 x 3.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-123-108 6 x 3.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-123-108 Supplier SeraCare™ Supplier No. 24000158
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For training, qualifying, and re-qualifying technical personnel in the performance of tests for the detection of antibodies to HIV-1, HIV-2 and HIV-1 group O, and HIV p24 antigen

  • Consists of six members manufactured from human serum or plasma that is negative for HBsAg and antibodies to HCV
  • With established reactivity in anti-HIV-1, anti-HIV-2, anti-HIV-1 group O, HIV p24 antigen and combination assays
  • Members formulated with various reactivities, two for antiHIV-1, one for anti-HIV-2, one for anti-HIV-1 group O, one for HIV p24 antigen and one nonreactive
  • Proclin™ (0.1%) added as preservative
  • Used with Abbott ARCHITECT HIV Ag/Ab and Abbott Prism HIV O Plus test kits
  • Also used as part of ongoing programs of lot acceptance and internal proficiency testing for these assays, to isolate system errors and in troubleshooting, as component of quality assurance programs
  • 1 vial per member, 3.5 mL per vial

Specifications

Content And Storage 2°C to 8°C
Format Panel
Type HIV 1/2/O/p24 Qualification Panel
Includes Six members with a range of reactivity for antibodies to HIV-1, HIV-2 and HIV-1 group O and for HIV p24 antigen
Intended Use Detection of Antibodies to HIV-1, HIV-2 and HIV-1 group O, and HIV p24 Antigen
Matrix Human Serum or Plasma
Quantity 6 x 3.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.