Learn More
Description
- Consists of seven positive members representing serial log dilutions of cultured HIV-1 type B 8E5 virus in HIV-1 RNA negative diluent and one negative member prepared from diluent
- Type B HIV-1 virus derived from 8E5 cells which containing intact but defective viral genome
- Diluent prepared from normal human plasma filtered through 0.2μm filter
- Sodium azide (0.09%) added as preservative
- Also used as part of operator training programs or for operator validation as part of ongoing laboratory quality assurance programs
- 1 Vial/member, 3.5mL/vial
Specifications
Specifications
| Content And Storage | -70°C |
| Format | Panel |
| Type | HIV-1 RNA Linearity Panel |
| Includes | Set of seven positive members representing serial log dilutions of cultured HIV-1 type B 8E5 virus in HIV-1 RNA negative diluent, one negative member prepared from the diluent |
| Intended Use | For optimizing, evaluating, monitoring and troubleshooting HIV-1 (Human Immunodeficiency Virus Type 1) RNA assays |
| Quantity | 8 x 3.5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.