Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ HIV-1 Seroconversion Panels

Catalog No. 22554557 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
4 x 1 mL
6 x 1 mL
10 x 1 mL
Type:
Bleed Dates April thru July 2008
Bleed Dates Feb/March 2000
Bleed Dates July/August 2001
Bleed Dates July/August 2007
Bleed Dates June 2008
Bleed Dates March/April 2010
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
22-554-557 4 x 1 mL Bleed Dates Feb/March 2000
22-554-549 10 x 1 mL Bleed Dates July/August 2007
22-554-550 10 x 1 mL Bleed Dates April thru July 2008
22-554-548 6 x 1 mL Bleed Dates July/August 2001
22-554-551 4 x 1 mL Bleed Dates June 2008
22-554-558 6 x 1 mL Bleed Dates March/April 2010
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22-554-557 Supplier SeraCare™ Supplier No. 06000253
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Undiluted samples collected from a single donor during development of infection and subsequent immune response

  • Maintain standards for assay validation, regulatory compliance, and method development
  • Each panel includes comprehensive, comparative data from a variety of methods and commercially available test kits, allowing user to independently challenge reagent, assay, and platform performance, sensitivity and specificity
  • Assembled from highly characterized, disease state patient samples, and developed specifically for infectious disease testing, both serology and NAT
  • Evaluate, monitor, and troubleshoot test and staff performance
  • Minimizes down time and accelerates verification of new test methods, reagent systems, and platforms
  • For research use only

Specifications

Content And Storage -65°C to -80°C
Type Bleed Dates Feb/March 2000
Intended Use Evaluating HIV-1 test system using well characterized specimens, to provide comprehensive data for comparative analysis
Quantity 4 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.