Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ HIV-1 Seroconversion Panels

Catalog No. 21123113 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
4 x 1.2 mL
5 x 1.2 mL
7 x 1.2 mL
Content And Storage:
-10°C
-65°C to -80°C
-70°C
Time Range:
11 Days in 2000
14 Days in 2002
15 Days in 2002
16 Days in 2000
33 Days in 2010
9 Days in 1999
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Content And Storage Time Range
21-123-113 4 x 1.2 mL -10°C 15 Days in 2002
21-123-106 7 x 1.2 mL -70°C 33 Days in 2010
21-123-109 4 x 1.2 mL -70°C 11 Days in 2000
21-123-112 4 x 1.2 mL -10°C 9 Days in 1999
21-123-111 5 x 1.2 mL -65°C to -80°C 14 Days in 2002
21-123-110 4 x 1.2 mL -70°C 16 Days in 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
These materials have not been treated and should be considered biohazardous. Some panel members were found positive by tests for HIV markers, including HIV RNA and anti-HIV.
Catalog No. 21-123-113 Supplier SeraCare™ Supplier No. 06000262
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These materials have not been treated and should be considered biohazardous. Some panel members were found positive by tests for HIV markers, including HIV RNA and anti-HIV.

For evaluating HIV-1 test systems using well-characterized specimens, and to provide comprehensive data for comparative analysis

  • Group of serial bleeds from an individual plasma donor during seroconversion
  • Contains set of undiluted plasma samples from single plasma donor
  • 1 vial per member, 1.2mL per vial

Cat. No. 21-123-109:

  • Four member panel
  • Collected over an 11-day period in 2000
  • Member 3 is a low titer HIV antigen sample, challenging for antigen and Ag/Ab tests
  • Member 4 is a challenging sample for antibody assays
Cat. No. 21-123-110:
  • Four member panel
  • Collected over an 16-day period in 2000
  • PRB974-02 is a moderate positive HIV RNA sample, challenging for HIV antigen assays
  • PRB974-03 is detected at very low signal to cutoff by all HIV Ag/Ab methods tested
  • PRB974-04 is a very challenging sample for anti-HIV assays
Cat. No. 21-123-111:
  • Five member panel
  • Collected over an 14-day period in 2002
  • PRB975-03 is a challenging sample for HIV RNA assays
  • Demonstrates improvement in sensitivity with introduction of “4th generation” HIV Ag/Ab tests
Cat. No. 21-123-112:
  • Four member panel
  • Collected over an 9-day period in 1999
  • Member 2 is a potentially challenging sample for HIV antigen and Ag/Ab tests
  • Demonstrates improvement in sensitivity with introduction of “4th generation” HIV Ag/Ab tests
Cat. No. 21-123-113:
  • Four member panel
  • Collected over an 15-day period in 2002
  • Member 1 is a challenging HIV RNA sample
  • Members 3 and 4 are both challenging for some antibody assays
Cat. No. 21-123-106:
  • Seven member panel
  • Collected over an 33-day period in 2010

Specifications

Content And Storage -10°C
Type HIV-1 Seroconversion Panel
Intended Use Evaluating HIV-1 test system using well characterized specimens, to provide comprehensive data for comparative analysis
Quantity 4 x 1.2 mL
Time Range 15 Days in 2002
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.