Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ QC Serology Qualification Panels

Catalog No. 22554091 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
5 x 4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-091 5 x 4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-554-091 Supplier SeraCare™ Supplier No. 24000140
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Detect testing problems before they affect laboratory operations

  • Convenient panels of characterized diluted samples designed to help testing laboratories:
    • Document personnel training
    • Troubleshoot assay performance
    • Evaluate proficiency
    • Document personnel training
    • Qualify incoming reagents
    • Fulfill regulatory requirements
    • Extensive array of panels helps your lab maintain the highest standards

Specifications

Content And Storage -70°C
Type AccuSet™ Qualification Panel
Intended Use Detection of HIV-1 RNA, HCV RNA, and HBV DNA
Quantity 5 x 4 mL
Source Human Serum or Plasma
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.