Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ Sensitivity and Linearity Panels

Catalog No. 22554283 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
15 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-283 15 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-554-283 Supplier SeraCare™ Supplier No. 08200183

missing translation for 'validMainframeMsg'

Specifications

Content And Storage -20°C
Type Lyme Mixed Titer Performance Panel
Includes Set of 15 aliquots with Anti-Borrelia burgdorferi reactivity ranging from negative to strongly positive
Intended Use Evaluating Anti-Borrelia burgdorferi tests with characterized samples, to provide comprehensive data for comparative analysis
Matrix Serum and Plasma
Quantity 15 x 0.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.