Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ West Nile Virus Seroconversion Panel

Catalog No. 22554295 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
5 x 1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-295 5 x 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-554-295 Supplier SeraCare™ Supplier No. 08200187
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Group of serial bleeds from an individual plasma donor during seroconversion for West Nile Virus (WNV) antibody IgM and WNV RNA

  • Intended for use by diagnostic manufacturers and clinical laboratorians to evaluate their WNV test systems using well-characterized specimens and to provide comprehensive data for comparative analysis
  • All units were maintained in a frozen state, except for the interval of dispensing into vials
  • No preservatives
  • For research use only: Not for use in diagnostic procedures

Specifications

Content And Storage -65°C to -80°C
Format Panel
Type West Nile Virus Seroconversion Panel
Includes Set of undiluted Plasma samples from a single Plasma donor collected during a period of WNV seroconversion
Intended Use Evaluating WNV test system with well characterized specimen, to provide comprehensive data for comparative analysis
Quantity 5 x 1.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.