Learn More
System Biosciences LLC SERAMIR EXOSOME RNA PURIFICATION KITSeraMir Exosome RNA Purification Column Kit (20 exoRNA columns) = 20 Preps
SDP
Catalog No. NC0946888
Encompass_Preferred
Efficiently purify RNA from already isolated exosomes with SBI’s most flexible SeraMir Kit.When you want to extract RNA from already isolated exosomes, choose the SeraMir Exosome RNA Column Purification Kit. With only SBI’s highly efficient SeraMir RNA purification columns and reagents, you can keep your exosome isolation and downstream RNA processing options open.
Catalog No. NC0946888 Supplier System Biosciences LLC Supplier No. RA808A1
Efficiently purify RNA from already isolated exosomes with SBI’s most flexible SeraMir Kit.When you want to extract RNA from already isolated exosomes, choose the SeraMir Exosome RNA Column Purification Kit. With only SBI’s highly efficient SeraMir RNA purification columns and reagents, you can keep your exosome isolation and downstream RNA processing options open.