Learn More
System Biosciences LLC SERAMIR EXOSOME AND RNA PURIFICATION KITSeraMir Exosome RNA Purification kit (5 mL ExoQuick and 20 exoRNA columns) = 20 Preps
SDP
Catalog No. NC0176181
Encompass_Preferred
Isolate exosomes and then purify exosomal RNAs for biomarker discovery and more (for serum, plasma, or ascites fluid). Optimized exosomal RNA extraction for biomarker discovery and more—for serum, plasma, and ascites fluid. Looking for biomarkers? Studying exosomal RNAs (exoRNAs)? Turn to SBI’s SeraMir Exosome RNA Purification Kit for optimized isolation of exoRNAs. The kit includes ExoQuick for efficient exosome isolation from serum, plasma, or ascites fluid, and SeraMir columns and reagents for purification of RNA from your isolated exosomes (for exoRNA isolation from other biofluids, try the SeraMir Exosome RNA Purification Kit for Media and Urine).
Catalog No. NC0176181 Supplier System Biosciences LLC Supplier No. RA806A1
Isolate exosomes and then purify exosomal RNAs for biomarker discovery and more (for serum, plasma, or ascites fluid). Optimized exosomal RNA extraction for biomarker discovery and more—for serum, plasma, and ascites fluid. Looking for biomarkers? Studying exosomal RNAs (exoRNAs)? Turn to SBI’s SeraMir Exosome RNA Purification Kit for optimized isolation of exoRNAs. The kit includes ExoQuick for efficient exosome isolation from serum, plasma, or ascites fluid, and SeraMir columns and reagents for purification of RNA from your isolated exosomes (for exoRNA isolation from other biofluids, try the SeraMir Exosome RNA Purification Kit for Media and Urine).