Learn More
Seracare Life Sciences Inc Seraseq ctDNA Complete Mutation Mix AF0.1% | 1x25ul
Catalog No. NC1717810
Encompass
To further accelerate the validation and clinical application of liquid biopsy-based ccfDNA-targeted NGS assays, SeraCare has developed the Seraseq ctDNA Complete that expands coverage of all relevant variant types including SNVs, INDELs, CNVs, and gene fusions in a single highly-multiplexed reference sample.
This item is not returnable. View return policy
Catalog No. NC1717810 Supplier Seracare Life Sciences Inc Supplier No. 07100532
To further accelerate the validation and clinical application of liquid biopsy-based ccfDNA-targeted NGS assays, SeraCare has developed the Seraseq ctDNA Complete that expands coverage of all relevant variant types including SNVs, INDELs, CNVs, and gene fusions in a single highly-multiplexed reference sample.