Learn More
Seracare Life Sciences Inc Seraseq ctDNA Complete Mutation Mix AF 0.5%, 25 uL
Catalog No. NC1717818,
Encompass
Expands coverage of all relevant variant types including SNVs, INDELs, CNVs, and gene fusions in a single highly-multiplexed reference sample.
This item is not returnable. View return policy
Catalog No. NC1717818 Supplier Seracare Life Sciences Inc Supplier No. 07100531
Expands coverage of all relevant variant types including SNVs, INDELs, CNVs, and gene fusions in a single highly-multiplexed reference sample.