Learn More
Seracare Life Sciences Inc Seraseq ctDNA Complete Mutation Mix AF 5%, 25 uL
Catalog No. NC1717822
Encompass
Expands coverage of all relevant variant types including SNVs, INDELs, CNVs, and gene fusions in a single highly-multiplexed reference sample.
This item is not returnable. View return policy
Catalog No. NC1717822 Supplier Seracare Life Sciences Inc Supplier No. 07100528
Expands coverage of all relevant variant types including SNVs, INDELs, CNVs, and gene fusions in a single highly-multiplexed reference sample.