Learn More
Seracare Life Sciences Inc Seraseq ctDNA Complete™ Mutation Mix AF1% | 1 x 25 µL
Catalog No. NC1870465
Encompass
To further accelerate the validation and clinical application of liquid biopsy-based ccfDNA-targeted NGS assays, SeraCare has developed the Seraseq ctDNA Complete that expands coverage of all relevant variant types including SNVs, INDELs, CNVs, and gene fusions in a single highly-multiplexed reference sample. These new products are offered in either a purified ctDNA format or a plasma-like matrix format, all precisely quantitated by digital PCR against a single well-characterized genomic background (GM24385), and orthogonally validated with NGS technology. This specific ctDNA Complete product has an allele frequency of 1%.
This item is not returnable. View return policy
Catalog No. NC1870465 Supplier Seracare Life Sciences Inc Supplier No. 07100530
To further accelerate the validation and clinical application of liquid biopsy-based ccfDNA-targeted NGS assays, SeraCare has developed the Seraseq ctDNA Complete that expands coverage of all relevant variant types including SNVs, INDELs, CNVs, and gene fusions in a single highly-multiplexed reference sample. These new products are offered in either a purified ctDNA format or a plasma-like matrix format, all precisely quantitated by digital PCR against a single well-characterized genomic background (GM24385), and orthogonally validated with NGS technology. This specific ctDNA Complete product has an allele frequency of 1%.