Learn More
Seracare Life Sciences Inc ctDNA Complete Mutation Mix WT (0%), 25 uL
Catalog No. NC1870464
Encompass
To further accelerate the validation and clinical application of liquid biopsy-based ccfDNA-targeted NGS assays, SeraCare has developed the Seraseq ctDNA Complete that expands coverage of all relevant variant types including SNVs, INDELs, CNVs, and gene fusions in a single highly-multiplexed reference sample. These new products are offered in either a purified ctDNA format or a plasma-like matrix format, all precisely quantitated by digital PCR against a single well-characterized genomic background (GM24385), and orthogonally validated with NGS technology.
This item is not returnable. View return policy
Catalog No. NC1870464 Supplier Seracare Life Sciences Inc Supplier No. 07100533
To further accelerate the validation and clinical application of liquid biopsy-based ccfDNA-targeted NGS assays, SeraCare has developed the Seraseq ctDNA Complete that expands coverage of all relevant variant types including SNVs, INDELs, CNVs, and gene fusions in a single highly-multiplexed reference sample. These new products are offered in either a purified ctDNA format or a plasma-like matrix format, all precisely quantitated by digital PCR against a single well-characterized genomic background (GM24385), and orthogonally validated with NGS technology.