Learn More
Seracare Life Sciences Inc Seraseq ctDNA Complete™ Reference Material AF5%, 5mL
Catalog No. NC1654538
Encompass
This specific ctDNA Complete product has an allele frequency of 5%. 25 unique multiplexed variants in 16 genes, covering 12 SNVs, 7 INDELs, 3 CNVs, and 3 SVs, in purified and plasma-like matrix formats. 6 different allele frequencies (AF) – WT, 0.1%, 0.5%, 1%, 2.5% and 5%. Allows end-to-end evaluation of assay performance across the entire workflow, including pre-analytic extraction steps. Variants quantitated with digital PCR and orthogonally validated by NGS. Blends with well-characterized GM24385 human genomic DNA as background ‘wild-type’ material. Manufactured within cGMP compliant, ISO 9001 and ISO 13485 certified facilities.
This item is not returnable. View return policy
Catalog No. NC1654538 Supplier Seracare Life Sciences Inc Supplier No. 07100669
This specific ctDNA Complete product has an allele frequency of 5%. 25 unique multiplexed variants in 16 genes, covering 12 SNVs, 7 INDELs, 3 CNVs, and 3 SVs, in purified and plasma-like matrix formats. 6 different allele frequencies (AF) – WT, 0.1%, 0.5%, 1%, 2.5% and 5%. Allows end-to-end evaluation of assay performance across the entire workflow, including pre-analytic extraction steps. Variants quantitated with digital PCR and orthogonally validated by NGS. Blends with well-characterized GM24385 human genomic DNA as background ‘wild-type’ material. Manufactured within cGMP compliant, ISO 9001 and ISO 13485 certified facilities.