Learn More
Seracare Life Sciences Inc Seraseq ctDNA Mutation Mix V4 AF5%, Highly Multiplexed Reference Material, 93 Clinically Relevant Variants, 25 µL
Catalog No. NC3314054
Encompass
Seraseq ctDNA Mutation Mix v4 AF5% is a refined reference material that accelerates validation of Next Generation Sequencing (NGS) assays for circulating tumor DNA. With 93 clinically relevant variants across 71 genes, it enables precise detection of low variant allele frequencies, reducing background noise and eliminating additional DNA isolation steps. Its formulation as purified DNA solution ensures readiness for NGS workflows, making it a reliable choice for clinical genomics.
Catalog No. NC3314054 Supplier Seracare Life Sciences Inc Supplier No. 07103100
Seraseq ctDNA Mutation Mix v4 AF5% is a refined reference material that accelerates validation of Next Generation Sequencing (NGS) assays for circulating tumor DNA. With 93 clinically relevant variants across 71 genes, it enables precise detection of low variant allele frequencies, reducing background noise and eliminating additional DNA isolation steps. Its formulation as purified DNA solution ensures readiness for NGS workflows, making it a reliable choice for clinical genomics.