Learn More
Seracare Life Sciences Inc Seraseq ctDNA Mutation Mix v2 WT | 1 x 25 µL
Catalog No. NC1894274
Encompass
To overcome the lack of patient-like reference materials for liquid biopsy, including drawbacks of existing methodologies such as sonication, SeraCare has developed a breakthrough technology that produces the most patient-like materials compared to native ctDNA. The Seraseq ctDNA Mutation Mix v2 is a highly multiplexed product for NGS-based ctDNA assays targeting cancer-relevant somatic mutations. This specific ctDNA Mutation Mix v2 product it the wild-type with an allele frequency of 0%.
This item is not returnable. View return policy
Catalog No. NC1894274 Supplier Seracare Life Sciences Inc Supplier No. 07100144
To overcome the lack of patient-like reference materials for liquid biopsy, including drawbacks of existing methodologies such as sonication, SeraCare has developed a breakthrough technology that produces the most patient-like materials compared to native ctDNA. The Seraseq ctDNA Mutation Mix v2 is a highly multiplexed product for NGS-based ctDNA assays targeting cancer-relevant somatic mutations. This specific ctDNA Mutation Mix v2 product it the wild-type with an allele frequency of 0%.