Learn More
Seracare Life Sciences Inc Seraseq FFPE TMB RM Score 9 | 2 x 10 µm
Catalog No. NC1894281
Encompass
To improve and harmonize the measurement of TMB, Seracare Life Sciences has generated and characterized tumor-normal human cell lines with different TMB scores for whole exome sequencing analysis of TMB in patient samples, for validating targeted NGS panel-based TMB assays, and as controls for TMB measurements in I-O clinical trial projects.
This item is not returnable. View return policy
Catalog No. NC1894281 Supplier Seracare Life Sciences Inc Supplier No. 07101308
To improve and harmonize the measurement of TMB, Seracare Life Sciences has generated and characterized tumor-normal human cell lines with different TMB scores for whole exome sequencing analysis of TMB in patient samples, for validating targeted NGS panel-based TMB assays, and as controls for TMB measurements in I-O clinical trial projects.