Learn More
Seracare Life Sciences Inc Seraseq Fusion RNA Mix v4 | 1 x 25 µL
Catalog No. NC1778303
Encompass
As diagnostic testing for chromosomal translocations and other complex structural variants continues to shift from single-site methods such as Fluorescent In Situ Hybridization (FISH) and targeted reverse transcription PCR (RT-PCR) to highly multiplexed next-generation sequencing (NGS) assays, there is an increasing need for highly multiplexed reference materials covering such mutations which may be rare or difficult to obtain. SeraCare continues to innovate to address this critical market need with an updated version designed for use with NGS assays that detect gene fusions via sequencing of RNA transcripts.
This item is not returnable. View return policy
Catalog No. NC1778303 Supplier Seracare Life Sciences Inc Supplier No. 07100497
As diagnostic testing for chromosomal translocations and other complex structural variants continues to shift from single-site methods such as Fluorescent In Situ Hybridization (FISH) and targeted reverse transcription PCR (RT-PCR) to highly multiplexed next-generation sequencing (NGS) assays, there is an increasing need for highly multiplexed reference materials covering such mutations which may be rare or difficult to obtain. SeraCare continues to innovate to address this critical market need with an updated version designed for use with NGS assays that detect gene fusions via sequencing of RNA transcripts.