Learn More
Seracare Life Sciences Inc Seraseq gDNA TMB Mix Score 26, 2 x 500 ng
Catalog No. NC1894283
Encompass
To improve and harmonize the measurement of TMB, we have generated and characterized tumor-normal human cell lines with different TMB scores by whole exome sequencing, for use in validating targeted NGS panel-based TMB assays, and as controls for TMB measurements in I-O clinical trial projects. The TMB reference materials are formated as genomic DNA (purified mix) and/or FFPE (require extraction).
This item is not returnable. View return policy
Catalog No. NC1894283 Supplier Seracare Life Sciences Inc Supplier No. 07101323
To improve and harmonize the measurement of TMB, we have generated and characterized tumor-normal human cell lines with different TMB scores by whole exome sequencing, for use in validating targeted NGS panel-based TMB assays, and as controls for TMB measurements in I-O clinical trial projects. The TMB reference materials are formated as genomic DNA (purified mix) and/or FFPE (require extraction).