Learn More
Seracare Life Sciences Inc Seraseq NTRK Fusion RNA Mix, Quality Reference Material, Targeted NGS Assays, 20 µL
Catalog No. NC2749767
Encompass_Preferred
The Seraseq NTRK Fusion RNA Mix is a high-quality reference material from LGC Clinical Diagnostics, designed for targeted NGS assays in cancer research. It enables the detection of RNA from 15 clinically relevant NTRK gene fusions, ensuring robust assay validation and control. Manufactured in a GMP-compliant facility, it provides consistent performance, streamlining library preparation and RNA detection in bioinformatics workflows.
Catalog No. NC2749767 Supplier Seracare Life Sciences Inc Supplier No. 07101696
The Seraseq NTRK Fusion RNA Mix is a high-quality reference material from LGC Clinical Diagnostics, designed for targeted NGS assays in cancer research. It enables the detection of RNA from 15 clinically relevant NTRK gene fusions, ensuring robust assay validation and control. Manufactured in a GMP-compliant facility, it provides consistent performance, streamlining library preparation and RNA detection in bioinformatics workflows.