Learn More
Seracare Life Sciences Inc Seraseq ctDNA Complete™ Reference Material AF0.1%, 1x5mL
Catalog No. NC1626188,
Encompass
To further accelerate the validation and clinical application of liquid biopsy-based ccfDNA targeted NGS assays, SeraCare has developed the Seraseq ctDNA Complete((TM) that expands coverage of all relevant variant types (SNVs, INDELs, CNVs, and gene fusions) in a single, highly multiplexed reference sample. These new products are offered in either a purified ctDNA format or a plasma-like matrix format, all precisely quantitated by digital PCR against a single well-characterized genomic background (GM24385), and orthogonally validated with NGS technology. This specific ctDNA Complete product has an allele frequency of 0.1%.
This item is not returnable. View return policy
Catalog No. NC1626188 Supplier Seracare Life Sciences Inc Supplier No. 07100673
To further accelerate the validation and clinical application of liquid biopsy-based ccfDNA targeted NGS assays, SeraCare has developed the Seraseq ctDNA Complete((TM) that expands coverage of all relevant variant types (SNVs, INDELs, CNVs, and gene fusions) in a single, highly multiplexed reference sample. These new products are offered in either a purified ctDNA format or a plasma-like matrix format, all precisely quantitated by digital PCR against a single well-characterized genomic background (GM24385), and orthogonally validated with NGS technology. This specific ctDNA Complete product has an allele frequency of 0.1%.