Learn More
Seracare Life Sciences Inc Seraseq Tri-Level Tumor Mutation DNA Mix v2 HC | 1 x 25 µL
Catalog No. NC1695536
Encompass
The Seraseq Tri-Level Tumor Mutation DNA Mix v2 (HC) is a multiplexed mixture of actionable biosynthetic DNA targets precisely blended with a single, well-characterized genomic background. This product is produced under rigorous design control and manufacturing practices, offered in a high concentration (HC) format at 5 ng/µL. Designed to assess the overall performance of your somatic mutation assay by next-generation sequencing (NGS), this unique product offers a range of allele frequencies in a single sample format, including single nucleotide variants (SNVs), insertion-deletion mutations (indels), and structural variants (SVs).
This item is not returnable. View return policy
Catalog No. NC1695536 Supplier Seracare Life Sciences Inc Supplier No. 07100097
The Seraseq Tri-Level Tumor Mutation DNA Mix v2 (HC) is a multiplexed mixture of actionable biosynthetic DNA targets precisely blended with a single, well-characterized genomic background. This product is produced under rigorous design control and manufacturing practices, offered in a high concentration (HC) format at 5 ng/µL. Designed to assess the overall performance of your somatic mutation assay by next-generation sequencing (NGS), this unique product offers a range of allele frequencies in a single sample format, including single nucleotide variants (SNVs), insertion-deletion mutations (indels), and structural variants (SVs).