Learn More
Seracare Life Sciences Inc Seraseq™ Tumor Mutation DNA Mix V2 AF10 HC, High-Concentration NGS Reference Material, 40 Unique DNA Targets, 25 µL
Catalog No. NC2029309
Encompass
The Seraseq™ Tumor Mutation DNA Mix v2 AF10 HC is a premium reference material engineered for Next-Generation Sequencing (NGS) assays. It features 40 unique biosynthetic DNA targets mixed with high-quality GM24385 human genomic DNA, at an allelic frequency of 10%. This product enhances the reliability of somatic mutation assays, ensuring optimal performance during library preparation and sequencing. Manufactured under stringent cGMP and ISO standards, it serves as a consistent solution for genomic analysis in disease research.
This item is not returnable. View return policy
Catalog No. NC2029309 Supplier Seracare Life Sciences Inc Supplier No. 07100094
The Seraseq™ Tumor Mutation DNA Mix v2 AF10 HC is a premium reference material engineered for Next-Generation Sequencing (NGS) assays. It features 40 unique biosynthetic DNA targets mixed with high-quality GM24385 human genomic DNA, at an allelic frequency of 10%. This product enhances the reliability of somatic mutation assays, ensuring optimal performance during library preparation and sequencing. Manufactured under stringent cGMP and ISO standards, it serves as a consistent solution for genomic analysis in disease research.