Learn More
Cayman Chemical Seratrodast, Purity: ≥98%, CAS: 112665-43-7, 10 mg
SDP
Catalog No. 502894690
Encompass_Preferred
A potent antagonist of the TXA2 receptor, blocking specific binding of U-46619 to platelets with an IC50 value of 7.4 nM and platelet aggregation induced by U-44069 with an IC50 value of 350 nM; commonly used to study the roles of the TP receptor in animal airways and in tissue samples
Catalog No. 50-289-4690 Supplier Cayman Chemical Supplier No. 900201410 MG
A potent antagonist of the TXA2 receptor, blocking specific binding of U-46619 to platelets with an IC50 value of 7.4 nM and platelet aggregation induced by U-44069 with an IC50 value of 350 nM; commonly used to study the roles of the TP receptor in animal airways and in tissue samples