Learn More
Selleck Chemical LLC Seratrodast(AA-2414) S2072-10mg
SDP
Catalog No. 503110898
Encompass_Preferred
Seratrodast (AA-2414 ABT-001) is a potent and selective thromboxane A2 receptor (TP) antagonist which is used primarily in the treatment of asthma and used as anti-inflammatory agent
Catalog No. 50-311-0898 Supplier Selleck Chemical LLC Supplier No. S207210MG
Seratrodast (AA-2414 ABT-001) is a potent and selective thromboxane A2 receptor (TP) antagonist which is used primarily in the treatment of asthma and used as anti-inflammatory agent