Learn More
Medchemexpress LLC Seratrodast | 112665-43-7 | 99.59% | 100 MG
SDP
Catalog No. 5000408254
Encompass_Preferred
Seratrodast (AA 2414) is an orally active antiasthmatic agent that functions as a thromboxane A2 receptor (TP) antagonist and a ferroptosis inhibitor. It reduces lipid ROS production, modulates the systemic xc-/GSH/GPX4 axis, and inhibits JNK phosphorylation and p53 expression. Seratrodast exhibits both anti-asthmatic and anti-epileptic activities.
  • Acts as a thromboxane A2 receptor (TP) antagonist
  • Functions as a ferroptosis inhibitor
  • Reduces lipid ROS production
  • Modulates the systemic xc-/GSH/GPX4 axis
  • Inhibits JNK phosphorylation and p53 expression
  • Exhibits anti-asthmatic activity
  • Exhibits anti-epileptic activity
Catalog No. 50-004-08254 Supplier Medchemexpress LLC Supplier No. HYB0774100MG
Seratrodast (AA 2414) is an orally active antiasthmatic agent that functions as a thromboxane A2 receptor (TP) antagonist and a ferroptosis inhibitor. It reduces lipid ROS production, modulates the systemic xc-/GSH/GPX4 axis, and inhibits JNK phosphorylation and p53 expression. Seratrodast exhibits both anti-asthmatic and anti-epileptic activities.
  • Acts as a thromboxane A2 receptor (TP) antagonist
  • Functions as a ferroptosis inhibitor
  • Reduces lipid ROS production
  • Modulates the systemic xc-/GSH/GPX4 axis
  • Inhibits JNK phosphorylation and p53 expression
  • Exhibits anti-asthmatic activity
  • Exhibits anti-epileptic activity