Learn More
Medchemexpress LLC Seratrodast | 112665-43-7 | 99.59% | 50 MG
SDP
Catalog No. 5000409122
Encompass_Preferred
Seratrodast (AA 2414) is an orally active antiasthmatic agent that acts as a thromboxane A2 receptor (TP) antagonist and ferroptosis inhibitor. It reduces lipid ROS production, modulates the systemic xc-/GSH/GPX4 axis, and inhibits JNK phosphorylation and p53 expression. It has demonstrated anti-asthmatic and anti-epileptic activity.
  • Acts as a thromboxane A2 receptor (TP) antagonist
  • Functions as a ferroptosis inhibitor
  • Reduces lipid ROS production
  • Modulates the systemic xc-/GSH/GPX4 axis
  • Inhibits JNK phosphorylation and p53 expression
  • Exhibits anti-asthmatic activity
  • Exhibits anti-epileptic activity
Catalog No. 50-004-09122 Supplier Medchemexpress LLC Supplier No. HYB077450MG
Seratrodast (AA 2414) is an orally active antiasthmatic agent that acts as a thromboxane A2 receptor (TP) antagonist and ferroptosis inhibitor. It reduces lipid ROS production, modulates the systemic xc-/GSH/GPX4 axis, and inhibits JNK phosphorylation and p53 expression. It has demonstrated anti-asthmatic and anti-epileptic activity.
  • Acts as a thromboxane A2 receptor (TP) antagonist
  • Functions as a ferroptosis inhibitor
  • Reduces lipid ROS production
  • Modulates the systemic xc-/GSH/GPX4 axis
  • Inhibits JNK phosphorylation and p53 expression
  • Exhibits anti-asthmatic activity
  • Exhibits anti-epileptic activity