Learn More
Medchemexpress LLC Seratrodast | 112665-43-7 | 99.6% | 1 ML
SDP
Catalog No. 5000408083
Encompass_Preferred
Seratrodast (AA 2414) is an orally active antiasthmatic agent. It functions as a thromboxane A2 receptor (TP) antagonist and a ferroptosis inhibitor, reducing lipid ROS production. This compound modulates the systemic xc-/GSH/GPX4 axis and inhibits JNK phosphorylation and p53 expression, demonstrating anti-asthmatic and anti-epileptic activities.
  • Acts as a thromboxane A2 receptor (TP) antagonist
  • Functions as a ferroptosis inhibitor
  • Reduces lipid ROS production
  • Modulates the systemic xc-/GSH/GPX4 axis
  • Inhibits JNK phosphorylation and p53 expression
  • Exhibits anti-asthmatic and anti-epileptic activity
Catalog No. 50-004-08083 Supplier Medchemexpress LLC Supplier No. HYB077410MM/1ML
Seratrodast (AA 2414) is an orally active antiasthmatic agent. It functions as a thromboxane A2 receptor (TP) antagonist and a ferroptosis inhibitor, reducing lipid ROS production. This compound modulates the systemic xc-/GSH/GPX4 axis and inhibits JNK phosphorylation and p53 expression, demonstrating anti-asthmatic and anti-epileptic activities.
  • Acts as a thromboxane A2 receptor (TP) antagonist
  • Functions as a ferroptosis inhibitor
  • Reduces lipid ROS production
  • Modulates the systemic xc-/GSH/GPX4 axis
  • Inhibits JNK phosphorylation and p53 expression
  • Exhibits anti-asthmatic and anti-epileptic activity