Learn More
Medchemexpress LLC Seratrodast | 112665-43-7 | 99.6% | 25 MG
SDP
Catalog No. 5000450716
Encompass_Preferred
Seratrodast (AA 2414) is an orally active antiasthmatic agent, a thromboxane A2 receptor (TP) antagonist, and a ferroptosis inhibitor. It reduces lipid ROS production, modulates the systemic xc-/GSH/GPX4 axis, and inhibits JNK phosphorylation and p53 expression. This compound exhibits anti-asthmatic and anti-epileptic activity.
  • Thromboxane A2 receptor (TP) antagonist
  • Ferroptosis inhibitor
  • Reduces lipid ROS production
  • Modulates the systemic xc-/GSH/GPX4 axis
  • Inhibits JNK phosphorylation and p53 expression
  • Exhibits anti-asthmatic activity
  • Exhibits anti-epileptic activity
  • For research use only
Catalog No. 50-004-50716 Supplier Medchemexpress LLC Supplier No. 25MGHYB0774
Seratrodast (AA 2414) is an orally active antiasthmatic agent, a thromboxane A2 receptor (TP) antagonist, and a ferroptosis inhibitor. It reduces lipid ROS production, modulates the systemic xc-/GSH/GPX4 axis, and inhibits JNK phosphorylation and p53 expression. This compound exhibits anti-asthmatic and anti-epileptic activity.
  • Thromboxane A2 receptor (TP) antagonist
  • Ferroptosis inhibitor
  • Reduces lipid ROS production
  • Modulates the systemic xc-/GSH/GPX4 axis
  • Inhibits JNK phosphorylation and p53 expression
  • Exhibits anti-asthmatic activity
  • Exhibits anti-epileptic activity
  • For research use only