Learn More
Medchemexpress LLC Seratrodast | 112665-43-7 | 99.6% | 5 MG
SDP
Catalog No. 5000409224
Encompass_Preferred
Seratrodast (AA 2414) is an orally active antiasthmatic agent that functions as a thromboxane A2 receptor (TP) antagonist and a ferroptosis inhibitor. It operates by reducing lipid reactive oxygen species (ROS) production, modulating the systemic xc-/GSH/GPX4 axis, and inhibiting JNK phosphorylation and p53 expression. This compound exhibits both anti-asthmatic and anti-epileptic activities.
  • Inhibits ferroptosis induced by RSL3 or Erastin in HT22 cells.
  • Down-regulates the phosphorylation level of JNK.
  • Inhibits ROS generation and lipid peroxidation induced by Erastin in HT22 cells.
  • Shows no significant cytotoxicity at concentrations below 10 μM in HT22 cells.
  • Prevents cell death in HT22 cells induced by ferroptosis inducers.
  • Inhibits neuronal ferroptosis in a pentylenetetrazole-induced mouse epilepsy model.
  • Reduces epileptic seizures in vivo.
Catalog No. 50-004-09224 Supplier Medchemexpress LLC Supplier No. HYB07745MG
Seratrodast (AA 2414) is an orally active antiasthmatic agent that functions as a thromboxane A2 receptor (TP) antagonist and a ferroptosis inhibitor. It operates by reducing lipid reactive oxygen species (ROS) production, modulating the systemic xc-/GSH/GPX4 axis, and inhibiting JNK phosphorylation and p53 expression. This compound exhibits both anti-asthmatic and anti-epileptic activities.
  • Inhibits ferroptosis induced by RSL3 or Erastin in HT22 cells.
  • Down-regulates the phosphorylation level of JNK.
  • Inhibits ROS generation and lipid peroxidation induced by Erastin in HT22 cells.
  • Shows no significant cytotoxicity at concentrations below 10 μM in HT22 cells.
  • Prevents cell death in HT22 cells induced by ferroptosis inducers.
  • Inhibits neuronal ferroptosis in a pentylenetetrazole-induced mouse epilepsy model.
  • Reduces epileptic seizures in vivo.