Learn More
Medchemexpress LLC Seratrodast (Standard) | 112665-43-7 | 98.2% | 25 MG
SDP
Catalog No. 5000450794
Encompass_Preferred
Seratrodast (Standard) is the analytical standard of Seratrodast, intended for research and analytical applications. It is an orally active antiasthmatic agent, a thromboxane A2 receptor (TP) antagonist, and a ferroptosis inhibitor.
  • Analytical standard for Seratrodast
  • Orally active antiasthmatic agent
  • Thromboxane A2 receptor (TP) antagonist
  • Ferroptosis inhibitor
  • Reduces lipid ROS production
  • Modulates the systemic xc-/GSH/GPX4 axis
  • Inhibits JNK phosphorylation and p53 expression
  • Exhibits anti-asthmatic activity
  • Exhibits anti-epileptic activity
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-50794 Supplier Medchemexpress LLC Supplier No. 25MGHYB0774R
Seratrodast (Standard) is the analytical standard of Seratrodast, intended for research and analytical applications. It is an orally active antiasthmatic agent, a thromboxane A2 receptor (TP) antagonist, and a ferroptosis inhibitor.
  • Analytical standard for Seratrodast
  • Orally active antiasthmatic agent
  • Thromboxane A2 receptor (TP) antagonist
  • Ferroptosis inhibitor
  • Reduces lipid ROS production
  • Modulates the systemic xc-/GSH/GPX4 axis
  • Inhibits JNK phosphorylation and p53 expression
  • Exhibits anti-asthmatic activity
  • Exhibits anti-epileptic activity
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS