Learn More
Medchemexpress LLC Seratrodast (Standard) | 112665-43-7 | 98.2% | 50 MG
SDP
Catalog No. 5000409434
Encompass_Preferred
Seratrodast (Standard) is the analytical standard for Seratrodast, designed for research and analytical applications. It functions as a thromboxane A2 receptor (TP) antagonist and a ferroptosis inhibitor, demonstrating anti-asthmatic and anti-epileptic properties.
  • Analytical standard for Seratrodast
  • Thromboxane A2 receptor (TP) antagonist
  • Ferroptosis inhibitor
  • Decreases lipid reactive oxygen species (ROS) production
  • Regulates the systemic xc-/GSH/GPX4 axis
  • Suppresses JNK phosphorylation and p53 expression
  • Exhibits anti-asthmatic and anti-epileptic activity
  • Commonly utilized in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-09434 Supplier Medchemexpress LLC Supplier No. HYB0774R50MG
Seratrodast (Standard) is the analytical standard for Seratrodast, designed for research and analytical applications. It functions as a thromboxane A2 receptor (TP) antagonist and a ferroptosis inhibitor, demonstrating anti-asthmatic and anti-epileptic properties.
  • Analytical standard for Seratrodast
  • Thromboxane A2 receptor (TP) antagonist
  • Ferroptosis inhibitor
  • Decreases lipid reactive oxygen species (ROS) production
  • Regulates the systemic xc-/GSH/GPX4 axis
  • Suppresses JNK phosphorylation and p53 expression
  • Exhibits anti-asthmatic and anti-epileptic activity
  • Commonly utilized in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS