Learn More
Aviva Systems Biology Corp SERBP1 Antibody - middle region (ARP40311_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40311_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SERBP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SERBP1. Alternate names: CGI-55, CHD3IP, DKFZp564M2423, FLJ90489, HABP4L, PAI-RBP1, PAIRBP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57388