Learn More
Medchemexpress LLC Serca2a activator 1 | 2139330-34-8 | 99.7% | 551.66 | C32H29N3O4S | 100MG
SDP
Catalog No. 5000236131
Encompass_Preferred
SERCA2a activator 1 is a small-molecule research compound that enhances the activity of the sarcoplasmic/endoplasmic reticulum Ca2+-ATPase isoform 2a (SERCA2a), reducing phospholamban inhibition and improving cardiac contractile relaxation in experimental systems. It is supplied as a light yellow to yellow solid and is also available as a 10 mM solution in DMSO for laboratory use; intended for research use only.
Catalog No. 50-002-36131 Supplier Medchemexpress LLC Supplier No. HY124873100MG
SERCA2a activator 1 is a small-molecule research compound that enhances the activity of the sarcoplasmic/endoplasmic reticulum Ca2+-ATPase isoform 2a (SERCA2a), reducing phospholamban inhibition and improving cardiac contractile relaxation in experimental systems. It is supplied as a light yellow to yellow solid and is also available as a 10 mM solution in DMSO for laboratory use; intended for research use only.