Learn More
TARGETMOL CHEMICALS INC SERCA2A ACTIVATOR 1 5MG
SDP
Catalog No. 502581141
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SERCA2a activator 1 is a sarco/endoplasmic reticulum Ca2+-dependent ATPase 2a activator. SERCA2a activator 1 decreases phospholamban inhibition and enhances the systolic and diastolic functions of the heart. purity: 99%
Catalog No. 50-258-1141 Supplier TARGETMOL CHEMICALS INC Supplier No. T168735MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SERCA2a activator 1 is a sarco/endoplasmic reticulum Ca2+-dependent ATPase 2a activator. SERCA2a activator 1 decreases phospholamban inhibition and enhances the systolic and diastolic functions of the heart. purity: 99%