Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SERCA2a activator 1 (compound A) | 2139330-34-8 | 99.7% | 551.66 g/mol | C32H29N3O4S | 50MG
SDP

Catalog No. 5000235353
Encompass_Preferred

SERCA2a activator 1 is a small-molecule activator of the sarco/endoplasmic reticulum Ca2+-dependent ATPase 2a (SERCA2a). It reduces phospholamban-mediated inhibition and can enhance cardiac contractile and relaxation responses in preclinical models. The compound is supplied for research use as a high-purity solid and is also available as a DMSO solution for assay use.

  • Activates SERCA2a to improve calcium handling in cardiac models.
  • High purity: 99.7%.
  • Molecular weight: 551.66 g/mol.
  • Chemical formula: C32H29N3O4S.
  • Physical form: light yellow to yellow solid powder.
  • Available as powder and as a 10 mM solution in DMSO.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months).

Catalog No. 50-002-35353 Supplier Medchemexpress LLC Supplier No. HY12487350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SERCA2a activator 1 is a small-molecule activator of the sarco/endoplasmic reticulum Ca2+-dependent ATPase 2a (SERCA2a). It reduces phospholamban-mediated inhibition and can enhance cardiac contractile and relaxation responses in preclinical models. The compound is supplied for research use as a high-purity solid and is also available as a DMSO solution for assay use.

  • Activates SERCA2a to improve calcium handling in cardiac models.
  • High purity: 99.7%.
  • Molecular weight: 551.66 g/mol.
  • Chemical formula: C32H29N3O4S.
  • Physical form: light yellow to yellow solid powder.
  • Available as powder and as a 10 mM solution in DMSO.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.