Learn More
Crescent Chemical Co Inc SERDOLIT CHELITE P
SDP
Catalog No. 50248553
Encompass_Preferred
SERDOLIT Chelite P analytical grade Off-white beads with a macroporous resin structure and a general affinity for polyvalent metal cations which may be employed for special techniques (e.g. 90Sr++ determination).Styrene-DVB matrix with aminomethylphosphonic acid grou
Catalog No. 50-248-553 Supplier Crescent Chemical Co Inc Supplier No. SE41706.01
SERDOLIT Chelite P analytical grade Off-white beads with a macroporous resin structure and a general affinity for polyvalent metal cations which may be employed for special techniques (e.g. 90Sr++ determination).Styrene-DVB matrix with aminomethylphosphonic acid grou