Learn More
Crescent Chemical Co Inc SERDOLIT MB-1
SDP
Supplier:  Crescent Chemical Co Inc SE40701.03
Encompass_Preferred
SERDOLIT MB-1 analytical grade Mixed-bed ion exchanger composed of a strongly acidic cation exchanger and a strongly basic anion exchanger (type I) in a ratio of 1:1.5 (v/v). Size - 500G Storage Conditions - +15 C TO +30 C Catalog Number - 40701
Catalog No. 50-248-498