Learn More
MSC ALPHATEC DISP COVERALL
Supplier:  MSC 00331140
Encompass
Disposable & Chemical Resistant Coveralls; Garment Style: Coveralls ; Size: 3X-Large ; Material: SMS ; Closure Type: 2-Way Zipper with Storm Flap ; Cuff Style: Elastic with Thumb-loop ; Ankle Style: Elastic
Catalog No. 50-273-9374