Learn More
MSC ALPHATEC DISP COVERALL
Supplier:  MSC 99799926
Encompass
Disposable & Chemical Resistant Coveralls; Garment Style: Coveralls ; Size: Medium ; Material: SMS ; Closure Type: 2-Way Zipper with Storm Flap; Finger Loops; Elastic ; Cuff Style: Elastic with Thumb-loop ; Ankle Style: Attached Anti-Skid Boots; Elastic
Catalog No. 50-273-6139