Learn More
Cayman Chemical Sergliflozin A, Purity: ≥98%, CAS: 360775-96-8, 5 mg
SDP
Catalog No. 502954003
Encompass_Preferred
An SGLT2 inhibitor (Ki = 2.39 nM for the human transporter) and active metabolite of sergliflozin etabonate; selective for SGLT2 over SGLT1 (Ki = 708 nM for the human transporter); co-infusion with glucose reduces maximal renal glucose reabsorption rates in rats
Catalog No. 50-295-4003 Supplier Cayman Chemical Supplier No. 143425 MG
An SGLT2 inhibitor (Ki = 2.39 nM for the human transporter) and active metabolite of sergliflozin etabonate; selective for SGLT2 over SGLT1 (Ki = 708 nM for the human transporter); co-infusion with glucose reduces maximal renal glucose reabsorption rates in rats