Learn More
TARGETMOL CHEMICALS INC SERGLIFLOZIN ETABONATE 5MG
SDP
Catalog No. 502580496
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sergliflozin etabonate (KGT-1251) is an inhibitor of SGLT2 and can be used in studies about type 2 diabetes and obesity. purity: 95%
Catalog No. 50-258-0496 Supplier TARGETMOL CHEMICALS INC Supplier No. T287575MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sergliflozin etabonate (KGT-1251) is an inhibitor of SGLT2 and can be used in studies about type 2 diabetes and obesity. purity: 95%