Learn More
OHAUS Serial Cable for Enviro-Genie
Catalog No. 01804419 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Cable
Catalog No. Product Type
01-804-419 Cable
1 options
Catalog No. 01-804-419 Supplier OHAUS Supplier No. 83041375
Add to Cart
Add to Cart
Serial Cable may be used to connect the Enviro-Genie to a computer via a serial port for data capture or programming.
  • It is a standard 9-pin, female to female straight through serial cable.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cable