Learn More
Ibidi USA Serial Connector for µ-Slides
SDP
Catalog No. NC1545505
Encompass
Serial Connectors connect two µ-Slides with Female Luers in a serial manner for flow experiments. polypropylene adapters and silicone tubing, autoclavable, sterile, 6x2 sets/box
This item is not returnable. View return policy
Catalog No. NC1545505 Supplier Ibidi USA Supplier No. 10830
Serial Connectors connect two µ-Slides with Female Luers in a serial manner for flow experiments. polypropylene adapters and silicone tubing, autoclavable, sterile, 6x2 sets/box