Learn More
BUCHI Serial Printer for M-560/M-565 Melting Point Instrument
SDP
Catalog No. 05405898 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Equipment):
M-560/M-565 Melting Point Instrument
Catalog No. For Use With (Equipment)
05-405-898 M-560/M-565 Melting Point Instrument
1 options
Catalog No. 05-405-898 Supplier BUCHI Supplier No. 11069766
Add to Cart
Add to Cart
Serial printer for maintaining documentation of calibration, melting and boiling points.
Achieve easy and fast determination of melting and boiling points for professional quality control with the Melting Point system and accessories.
  • Printer for GLP/GMP-compliant documentation of calibration results and melting and boiling point determination
  • Speed up input of sample parameters with the optional keyboard
  • Ensure consistent sample preparation using compaction wire
Specifications
For Use With (Application) Maintaining Documentation of Results
For Use With (Equipment) M-560/M-565 Melting Point Instrument
Model Star SP712