Learn More
Molecular Devices Molecular Devices Reader Cable, MAC-Series 1, Emax and Vmax Compatible
Catalog No. 5000014444
Encompass_Preferred
Molecular Devices replacement cable designed to connect MAC-Series 1 readers to compatible Emax and Vmax microplate readers for instrument control and data transfer.
  • Replacement interconnect cable for legacy microplate readers.
  • Enables data and control signal transfer between instruments.
  • Plug-and-play design for straightforward installation.
  • Supplied as a single unit for spare parts inventory.
Catalog No. 50-000-14444 Supplier Molecular Devices Supplier No. 90000077
Molecular Devices replacement cable designed to connect MAC-Series 1 readers to compatible Emax and Vmax microplate readers for instrument control and data transfer.
  • Replacement interconnect cable for legacy microplate readers.
  • Enables data and control signal transfer between instruments.
  • Plug-and-play design for straightforward installation.
  • Supplied as a single unit for spare parts inventory.