Learn More
AdipoGen Sericic acid (25 mg)
SDP
Supplier:  AdipoGen AGCN20402M025
Encompass_Preferred
Chemical. CAS: 55306-03-1. Formula: C30H48O6. MW: 504.7. Isolated from Vochysia divergens. Anti-inflammatory. Antibacterial. Antifungal. Haemolytic agent. Weak phosphodiesterase 4 (PDE4) inhibitor. Antioxidant.
Catalog No. 50-205-3573